CAS RN 42757-55-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Benzene,1,1'-sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)-] (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzene,1,1'-sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)-] (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzene,1,1'-sulfonylbis[3,5-dibromo-4-(2,3-dibromopropoxy)-] (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.