CAS RN 42417-65-2
IUPAC Navn:
2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Synonymer:
2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Molekylvægt (g/mol):
265.31
Molekylformel:
C14H19NO4
InChi Key:
NNEHOKZDWLJKHP-UHFFFAOYNA-N
SMILES:
CC(C)C(N(C)C(=O)OCC1=CC=CC=C1)C(O)=O
1
–
3
af
3
Resultater
1
(S)-N-(Benzyloxycarbonyl)-N-methylvaline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-N-(Benzyloxycarbonyl)-N-methylvaline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-N-(Benzyloxycarbonyl)-N-methylvaline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.