CAS RN 401564-34-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-Des((3-methyl-1-phenyl-1H-pyrazol-5-yl)-1-piperazinyl)-4-hydroxy-N-(tert-butyl formate) Teneligliptin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Des((3-methyl-1-phenyl-1H-pyrazol-5-yl)-1-piperazinyl)-4-hydroxy-N-(tert-butyl formate) Teneligliptin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Des((3-methyl-1-phenyl-1H-pyrazol-5-yl)-1-piperazinyl)-4-hydroxy-N-(tert-butyl formate) Teneligliptin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.