CAS RN 40150-98-9
IUPAC Navn:
4-(2-methylpropyl)benzaldehyde
Synonymer:
4-(2-methylpropyl)benzaldehyde
Molekylvægt (g/mol):
162.23
Molekylformel:
C11H14O
InChi Key:
LXPWGAZYJHUWPM-UHFFFAOYSA-N
SMILES:
CC(C)CC1=CC=C(C=O)C=C1
1
–
3
af
3
Resultater
1
4-(2-Methylpropyl)benzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(2-Methylpropyl)benzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(2-Methylpropyl)benzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.