CAS RN 388633-49-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
6-Methyl-2-(4-methylphenyl)-Alpha-oxo-imidazo[1,2-a]pyridine-3-acetic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Methyl-2-(4-methylphenyl)-Alpha-oxo-imidazo[1,2-a]pyridine-3-acetic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.