CAS RN 36701-89-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Diflufenican metabolite AE B107137 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
2-[3-(Trifluoromethyl)phenoxy]nicotinic Acid, TRC
CAS: 36701-89-0 Molekylær formel: C13 H8 F3 N O3 Molekylvægt (g/mol): 283.2 Synonym: Diflufenican Metabolite AE B107137,2-[3-(Trifluoromethyl)phenoxy]-3-pyridinecarboxylic acid,2-(3-Trifluoromethylphenoxy)-3-pyridinecarboxylic acid,2-(3-Trifluoromethylphenoxy)nicotinic acid,3-Pyridinecarboxylic acid, 2-[3-(trifluoromethyl)phenoxy]- IUPAC navn: 2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxylic acid SMIL: OC(=O)c1cccnc1Oc2cccc(c2)C(F)(F)F