Få mere at vide
2-[3-(Trifluoromethyl)phenoxy]nicotinic Acid, TRC
Artikelnummer. 30138404
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138404 10 mg
30133247 100 mg
30010566 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138404 Leverandør Toronto Research Chemicals Leverandørnr. TRCT90177810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 36701-89-0
Molekylær formel C13 H8 F3 N O3
Molekylvægt (g/mol) 283.2
Synonym Diflufenican Metabolite AE B107137, 2-[3-(Trifluoromethyl)phenoxy]-3-pyridinecarboxylic acid, 2-(3-Trifluoromethylphenoxy)-3-pyridinecarboxylic acid, 2-(3-Trifluoromethylphenoxy)nicotinic acid, 3-Pyridinecarboxylic acid, 2-[3-(trifluoromethyl)phenoxy]-
IUPAC navn 2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxylic acid
SMIL OC(=O)c1cccnc1Oc2cccc(c2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Diflufenican Metabolite AE B107137
Mængde 10 mg
InChI formel InChI=1S/C13H8F3NO3/c14-13(15,16)8-3-1-4-9(7-8)20-11-10(12(18)19)5-2-6-17-11/h1-7H,(H,18,19)
Formel vægt 283.0456