CAS RN 3599-89-1
IUPAC Navn:
propanimidamide hydrochloridyl
Synonymer:
propamidine hydrochloridyl
Molekylvægt (g/mol):
107.56
Molekylformel:
C3H8ClN2
InChi Key:
KBOZRAZQYBTUJV-UHFFFAOYSA-N
SMILES:
[Cl].CCC(N)=N
1
–
3
af
3
Resultater
1
Propioamidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Propioamidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Propioamidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.