CAS RN 359436-61-6
IUPAC Navn:
3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanehydrazide; trifluoroacetic acid
Synonymer:
3-(2,5-dioxopyrrol-1-yl)propanehydrazide; trifluoroacetic acid
Molekylvægt (g/mol):
297.19
Molekylformel:
C9H10F3N3O5
InChi Key:
BADCXPKRBUEEMA-UHFFFAOYSA-N
SMILES:
OC(=O)C(F)(F)F.NNC(=O)CCN1C(=O)C=CC1=O
1
–
3
af
3
Resultater
1
3-Maleimidopropionic Acid Hydrazonium Trifluoroacetic Acid Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Maleimidopropionic Acid Hydrazonium Trifluoroacetic Acid Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Maleimidopropionic Acid Hydrazonium Trifluoroacetic Acid Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.