Få mere at vide
3-Maleimidopropionic Acid Hydrazonium Trifluoroacetic Acid Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Maleimidopropionic Acid Hydrazonium, Trifluoroacetate |
| CAS | 359436-61-6 |
| Molekylær formel | C7 H9 N3 O3 . C2 H F3 O2 |
| Mængde | 10 mg |
| Synonym | 1H-Pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo-, hydrazide 2,2,2-trifluoroacetate (1:1), 1H-Pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo-, hydrazide, mono(trifluoroacetate) (9CI), N-(β-Maleimidopropionic acid)hydrazide trifluoroacetate, 2,5-Dioxo-1H-pyrrole-1-propanoic acid hydrazide trifluoroacetate, 3-Maleimidopropiohydrazide trifluoroacetate, 3-Maleimidopropionic acid hydrazonium trifluoroacetate, 3-Maleimidopropionic acid hydrazonium trifluoroacetic acid salt |
| InChI formel | InChI=1S/C7H9N3O3.C2HF3O2/c8-9-5(11)3-4-10-6(12)1-2-7(10)13;3-2(4,5)1(6)7/h1-2H,3-4,8H2,(H,9,11);(H,6,7) |
| SMIL | NNC(=O)CCN1C(=O)C=CC1=O.OC(=O)C(F)(F)F |
| IUPAC navn | 3-(2,5-dioxopyrrol-1-yl)propanehydrazide;2,2,2-trifluoroacetic acid |
| Molekylvægt (g/mol) | 297.19 |
| Formel vægt | 297.0573 |