CAS RN 35943-35-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ API-2
Selective inhibitor of Akt/PKB signaling. Antitumor and antiviral
2
Triciribine, TRC
CAS: 35943-35-2 Molekylær formel: C13 H16 N6 O4 Molekylvægt (g/mol): 320.3 Synonym: 1,5-Dihydro-5-methyl-1-β-D-ribofuranosyl-1,4,5,6,8-pentaazaacenaphthylen-3-amine SMIL: CN1N=C(N)c2cn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c4ncnc1c24
3
Triciribine, MedChemExpress
MedChemExpress Triciribine is a DNA synthesis inhibitor, also inhibits Akt and HIV-1/2 with IC50 of 130 nM, and 0.02-0.46 μM, respectively.