Få mere at vide
Triciribine, MedChemExpress
Artikelnummer. 16499117
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16499117 5 mg -
16482627 50 mg -
16456538 10 mg -
16446776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16499117 Leverandør Medchem Express Leverandørnr. HY15457
MedChemExpress Triciribine is a DNA synthesis inhibitor, also inhibits Akt and HIV-1/2 with IC50 of 130 nM, and 0.02-0.46 μM, respectively.
Triciribine is a DNA synthesis inhibitor, also inhibits Akt and HIV-1/2 with IC50 of 130 nM, and 0.02-0.46 μM, respectively.
Tekniske data
Kemisk navn eller materiale Triciribine
CAS 35943-35-2
Farve Earth Yellow
Renhedsgrad noter Research
Molekylær formel C13H16N6O4
Mængde 5 mg
Synonym API-2 NSC 154020 TCN
Opløselighedsinformation DMSO : 100 mg/mL (312.21 mM; Need ultrasonic)
SMIL OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C3=NC=NC4=C3C(C(N)=NN4C)=C2
Molekylvægt (g/mol) 320.3
Formel vægt 320.3
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease