CAS RN 3530-19-6
IUPAC Navn:
disodium (8Z)-8-(2-{4'-[2-(4-ethoxyphenyl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-7-oxo-7,8-dihydronaphthalene-1,3-disulfonate
Synonymer:
disodium (8Z)-8-(2-{4'-[2-(4-ethoxyphenyl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}hydrazin-1-ylidene)-7-oxonaphthalene-1,3-disulfonate
Molekylvægt (g/mol):
676.63
Molekylformel:
C30H22N4Na2O8S2
InChi Key:
YFRKTOTWUMQDNQ-NQLNAIKTSA-L
SMILES:
[Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(C=C1)C1=CC=C(N\N=C2/C(=O)C=CC3=CC(=CC(=C23)S([O-])(=O)=O)S([O-])(=O)=O)C=C1
1
–
3
af
3
Resultater
1
Direct Scarlet B (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Direct Scarlet B (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Direct Scarlet B (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.