Få mere at vide
Direct Scarlet B (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Direct Scarlet B (Technical Grade) |
| CAS | 3530-19-6 |
| Molekylær formel | C30H22N4Na2O8S2 |
| Mængde | 2.5 g |
| Synonym | 8-[2-[4'-[2-(4-ethoxyphenyl)diazenyl][1,1'-biphenyl]-4-yl]diazenyl]-7-hydroxy-1,3-Naphthalenedisulfonic acid sodium salt, 8-[[4’-[(4-ethoxyphenyl)azo][1,1'-biphenyl]-4-yl]azo]-7-hydroxy-1,3-Naphthalenedisulfonic acid disodium salt, C.I. Direct Red 37, disodium salt (8CI), Airedale Scarlet BD, Amanil Fast Scarlet B, Atlantic Scarlet B, Azine Scarlet BX, Benzanil Scarlet B, Benzanol Brilliant Scarlet B, Benzo Scarlet B, Benzyl Scarlet BS, C.I. 22240, C.I. Direct Red 37, Calcomine Scarlet B, Chloramine Red B, Chrome Leather Scarlet BS, Diamine Scarlet BA-CF, Diaphtamine Fast Red B, Diazol Scarlet B, Diphenyl Red BS, Diphenyl Scarlet BS, Direct Fast Scarlet B, Direct Fast Scarlet BS, Direct Scarlet BS, Erie Scarlet B, Fenamin Scarlet B, Kayaku Direct Scarlet B, Kayaku Direct Scarlet B New, Mitsui Direct Scarlet BS, Nippon Scarlet B, Nitto Direct Scarlet B, Paper Scarlet BAP, Paramine Fast Scarlet B, Phenamine Scarlet B, Pontamine Scarlet B, Shikiso Direct Scarlet BX, Tertrodirect Ponceau B, Triazol Fast Scarlet B, Union Fast Scarlet B |
| InChI formel | InChI=1S/C30H24N4O8S2.2Na/c1-2-42-25-14-12-24(13-15-25)32-31-22-8-3-19(4-9-22)20-5-10-23(11-6-20)33-34-30-27(35)16-7-21-17-26(43(36,37)38)18-28(29(21)30)44(39,40)41;;/h3-18,35H,2H2,1H3,(H,36,37,38)(H,39,40,41);;/q;2*+1/p-2 |
| SMIL | [Na+].[Na+].CCOc1ccc(cc1)N=Nc2ccc(cc2)c3ccc(cc3)N=Nc4c(O)ccc5cc(cc(c45)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC navn | disodium;8-[(E)-[4-[4-[(E)-(4-ethoxyphenyl)diazenyl]phenyl]phenyl]diazenyl]-7-hydroxynaphthalene-1,3-disulfonate |
| Molekylvægt (g/mol) | 676.63 |
| Formel vægt | 676.0674 |