CAS RN 35212-22-7
IUPAC Navn:
3-phenyl-7-(propan-2-yloxy)-4H-chromen-4-one
Synonymer:
osten quinogin osteoquine ipriflavona ipriflavone ipriflavonum yambolap
Molekylvægt (g/mol):
280.32
Molekylformel:
C18H16O3
InChi Key:
SFBODOKJTYAUCM-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=C2C(=O)C(=COC2=C1)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
Ipriflavone, MedChemExpress
MedChemExpress Ipriflavone is a synthetic isoflavone derivative used to suppress bone resorption.