CAS RN 335161-03-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
AM-694 (1-(5-Fluoropentyl)-3-(2-iodobenzoyl)indole) 0.1 mg/ml in Acetonitrile, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
AM-694 (1-(5-Fluoropentyl)-3-(2-iodobenzoyl)indole) 1.0 mg/ml in Acetonitrile, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.