CAS RN 33458-93-4
IUPAC Navn:
9-oxo-6-(propan-2-yloxy)-9H-xanthene-2-carboxylic acid
Synonymer:
6-isopropoxy-9-oxoxanthene-2-carboxylic acid
Molekylvægt (g/mol):
298.29
Molekylformel:
C17H14O5
InChi Key:
AQFFXPQJLZFABJ-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=C2C(OC3=CC=C(C=C3C2=O)C(O)=O)=C1
1
–
1
af
1
Resultater
1
AH 6809, MedChemExpress
MedChemExpress AH 6809 is an antagonist of EP and DP receptor, with Kis of 1217, 1150, 1597, and 1415 nM for the cloned human EP1, EP2, EP3-III, and DP receptor respectively. AH 6809 has a Ki of 350 nM for mouse EP2 receptor.