CAS RN 331963-29-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
I2906, MedChemExpress
MedChemExpress I2906 showed antimycobacterial and cytotoxic activity against mycobacterium tuberculosis.