CAS RN 33045-52-2
IUPAC Navn:
methyl 2-methoxy-5-sulfamoylbenzoate
Synonymer:
methyl 2-methoxy-5-sulfamoylbenzoate
Molekylvægt (g/mol):
245.25
Molekylformel:
C9H11NO5S
InChi Key:
MKDYDRQLKPGNNU-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O
1
–
4
af
4
Resultater
1
5-(Aminosulfonyl)-2-methoxy-benzoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sulpiride EP Impurity B, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
5-(Aminosulfonyl)-2-methoxy-benzoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
5-(Aminosulfonyl)-2-methoxy-benzoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.