CAS RN 3278-31-7
IUPAC Navn:
1-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonymer:
1-[4-(2,5-dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione
Molekylvægt (g/mol):
268.23
Molekylformel:
C14H8N2O4
InChi Key:
AQGZJQNZNONGKY-UHFFFAOYSA-N
SMILES:
O=C1C=CC(=O)N1C1=CC=C(C=C1)N1C(=O)C=CC1=O
1
–
3
af
3
Resultater
1
1,4-Phenylene-bis-maleimide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,4-Phenylene-bis-maleimide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,4-Phenylene-bis-maleimide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.