CAS RN 307986-98-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ NNC 0640
Negative allosteric modulator of glucagon and GLP-1 receptors
2
NNC-0640, MedChemExpress
MedChemExpress NNC-0640 is a potent human G-protein-coupled glucagon receptor (GCGR) negative allosteric modulator (NAM) with an IC50 of 69.2 nM.