CAS RN 30163-20-3
IUPAC Navn:
2-amino-3-(3-bromophenyl)propanoic acid
Synonymer:
2-amino-3-(3-bromophenyl)propanoic acid
Molekylvægt (g/mol):
244.09
Molekylformel:
C9H10BrNO2
InChi Key:
GDMOHOYNMWWBAU-UHFFFAOYNA-N
SMILES:
NC(CC1=CC(Br)=CC=C1)C(O)=O
1
–
2
af
2
Resultater
1
DL-3-Bromophenylalanine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Bromo-DL-phenylalanine, 95%, Thermo Scientific™
CAS: 30163-20-3 Molekylær formel: C9H10BrNO2 Molekylvægt (g/mol): 244.088 MDL nummer: MFCD05227935 InChI nøgle: GDMOHOYNMWWBAU-UHFFFAOYSA-N Synonym: 2-amino-3-3-bromophenyl propanoic acid,3-bromo-dl-phenylalanine,3-bromophenylalanine,dl-3-br-phe-oh,h-dl-phe 3-br-oh,dl-3-bromophenylalanine,pubchem18643,3-bromophenylalanine #,2-3-bromobenzyl glycine,dl-3-br-phe PubChem CID: 554078 IUPAC navn: 2-amino-3-(3-bromophenyl)propanoic acid SMIL: C1=CC(=CC(=C1)Br)CC(C(=O)O)N