CAS RN 299433-10-6
IUPAC Navn:
1-(naphthalene-1-sulfonyl)-6-{octahydropyrrolo[1,2-a]pyrazin-2-yl}-1H-indole dihydrochloride
Synonymer:
6-{hexahydro-1H-pyrrolo[1,2-a]pyrazin-2-yl}-1-(naphthalene-1-sulfonyl)indole dihydrochloride
Molekylvægt (g/mol):
504.47
Molekylformel:
C25H27Cl2N3O2S
InChi Key:
XQQYORJJFZQWNA-UHFFFAOYNA-N
SMILES:
Cl.Cl.O=S(=O)(N1C=CC2=CC=C(C=C12)N1CCN2CCCC2C1)C1=C2C=CC=CC2=CC=C1
1
–
1
af
1
Resultater
1
NPS ALX Compound 4a, MedChemExpress
MedChemExpress NPS ALX Compound 4a is a potent and selective 5-hydroxytryptamine6 (5-HT6) receptor antagonist, with an IC50 of 7.2 nM and a Ki of 0.2 nM.