CAS RN 2969-13-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-[4-[3-[(EZ)-2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]piperazin-1-yl]ethyl Acetate Dihydrochloride (O-Acetylflupentixol Dihydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.