Få mere at vide
2-[4-[3-[(EZ)-2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]piperazin-1-yl]ethyl Acetate Dihydrochloride (O-Acetylflupentixol Dihydrochloride), Mikromol™
Beskrivelse
2-[4-[3-[(EZ)-2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]piperazin-1-yl]ethyl Acetate Dihydrochloride (O-Acetylflupentixol Dihydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | O-Acetylflupentixol Dihydrochloride |
| CAS | 2969-13-3 |
| Analyt- eller komponentnavne | O-Acetylflupentixol Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H27 F3 N2 O2 S . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]-, 1-acetate, hydrochloride (1:2), 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)thioxanthen-9-ylidene]propyl]-, acetate (ester), dihydrochloride (8CI), 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)thioxanthen-9-ylidene]propyl]-, acetate, dihydrochloride (7CI), Flupentixol Dihydrochloride Imp. E (EP) as Dihydrochloride, Flupentixol Dihydrochloride Impurity E as Dihydrochloride, Flupentixol Impurity E as Dihydrochloride |
| InChI formel | InChI=1S/C25H27F3N2O2S.2ClH/c1-18(31)32-16-15-30-13-11-29(12-14-30)10-4-6-20-21-5-2-3-7-23(21)33-24-9-8-19(17-22(20)24)25(26,27)28;;/h2-3,5-9,17H,4,10-16H2,1H3;2*1H |
| SMIL | Cl.Cl.CC(=O)OCCN1CCN(CCC=C2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC navn | 2-[4-[3-[2-(trifluoromethyl)thioxanthen-9-ylidene]propyl]piperazin-1-yl]ethyl acetate;dihydrochloride |
| Vis mere |