CAS RN 2953-50-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
cis-1,3,3-Trichloro-1-propene (cis-trans mixture, ~70% Z), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
cis-1,3,3-Trichloro-1-propene (cis-trans mixture, ~70% Z), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
cis-1,3,3-Trichloro-1-propene (cis-trans mixture, ~70% Z), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.