CAS RN 259525-01-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Enecadin, MedChemExpress
MedChemExpress Enecadin is a neuroprotective agent extracted from patent US 8623823 B2.