CAS RN 2408611-30-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-[(1S,3R)-3-Hydroxycyclopentyl]-9-methoxy-1,8-dioxo-N-[(2,4,6-trifluorophenyl)methyl]pyrido[1,2-a]pyrazine-7-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[(1S,3R)-3-Hydroxycyclopentyl]-9-methoxy-1,8-dioxo-N-[(2,4,6-trifluorophenyl)methyl]pyrido[1,2-a]pyrazine-7-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.