CAS RN 23731-38-6
IUPAC Navn:
methyl (²H)formate
Synonymer:
methyl (²H)formate
Molekylvægt (g/mol):
61.06
Molekylformel:
C2H4O2
InChi Key:
TZIHFWKZFHZASV-VMNATFBRSA-N
SMILES:
[2H]C(=O)OC
1
–
4
af
4
Resultater
1
Methyl Formate-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl Formate-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl Formate-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Methyl Formate-d1, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.