Få mere at vide
Methyl Formate-d1, CDN
Artikelnummer. 30321308
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30321308 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321308 Leverandør CDN Leverandørnr. CDND00695G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl Formate-d
CAS 23731-38-6
Molekylær formel C2 D H3 O2
Mængde 5 g
Synonym Formic-d acid, methyl ester (6 CI, 7 CI, 8 CI, 9 CI), Methyl deuterioformate, Methyl Formate-d1, Methyl Formate-D, Formic-d acid methyl ester, Formic acid methyl ester-d1
InChI formel InChI=1 S/C2H4O2/c1-4-2-3/h2H,1H3/i2D
SMIL [2 H]C(=O)OC
IUPAC navn methyl deuterioformate
Molekylvægt (g/mol) 61.0581
Formel vægt 61.0274 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity