CAS RN 23277-43-2
IUPAC Navn:
(1S,5R,13R,14S,17S)-4-(cyclobutylmethyl)-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-triene-10,14,17-triol hydrochloride
Synonymer:
nalbuphine hydrochloride
Molekylvægt (g/mol):
393.91
Molekylformel:
C21H28ClNO4
InChi Key:
YZLZPSJXMWGIFH-BCXQGASESA-N
SMILES:
Cl.O[C@H]1CC[C@@]2(O)[C@H]3CC4=CC=C(O)C5=C4[C@@]2(CCN3CC2CCC2)[C@H]1O5
1
–
2
af
2
Resultater
1
Nalbuphine Hydrochloride, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Nalbuphine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.