Få mere at vide
Nalbuphine Hydrochloride, LoGiCal
Artikelnummer. 30460560
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460560 10 mg
1 options
Artikelnummer. 30460560 Leverandør LoGiCal Leverandørnr. LGCFOR0341.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nalbuphine Hydrochloride
CAS 23277-43-2
Analyt- eller komponentnavne Nalbuphine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C21 H27 N O4 . Cl H
Certificeringer/compliance ISO 9001
Synonym Nalbuphine Hydrochloride
InChI formel InChI=1S/C21H27NO4.ClH/c23-14-5-4-13-10-16-21(25)7-6-15(24)19-20(21,17(13)18(14)26-19)8-9-22(16)11-12-2-1-3-12;/h4-5,12,15-16,19,23-25H,1-3,6-11H2;1H/t15-,16+,19-,20-,21+;/m0./s1
Løsningstype Neat
SMIL Cl.O[C@H]1CC[C@@]2(O)[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3CC6CCC6)c45
IUPAC navn (4R,4aS,7S,7aR,12bS)-3-(cyclobutylmethyl)-1,2,4,5,6,7,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-4a,7,9-triol;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 393.9
Mængde 10 mg
Formel vægt 393.1707 g/mol