CAS RN 2309668-44-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Derazantinib Racemate, MedChemExpress
MedChemExpress Derazantinib Racemate (ARQ-087 Racemate) is the racemate of Derazantinib. Derazantinib is an orally bioavailable, ATP competitive tyrosine kinase inhibitor; exhibits potent activity against FGFR1-3 chondrocytes with IC50s of 4.5, 1.8, and 4.5 nM, respectively.