Få mere at vide
Derazantinib Racemate, MedChemExpress
Artikelnummer. 16440336
Encompass_Preferred
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
16440336 2 mg
1 options
Artikelnummer. 16440336 Leverandør Medchem Express Leverandørnr. HY19981A
MedChemExpress Derazantinib Racemate (ARQ-087 Racemate) is the racemate of Derazantinib. Derazantinib is an orally bioavailable, ATP competitive tyrosine kinase inhibitor; exhibits potent activity against FGFR1-3 chondrocytes with IC50s of 4.5, 1.8, and 4.5 nM, respectively.
Derazantinib Racemate (ARQ-087 Racemate) is the racemate of Derazantinib. Derazantinib is an orally bioavailable, ATP competitive tyrosine kinase inhibitor
Tekniske data
Kemisk navn eller materiale Derazantinib Racemate
CAS 2309668-44-6
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C29H29FN4O
Mængde 2 mg
Synonym ARQ-087 Racemate
Opløselighedsinformation DMSO : 100 mg/mL (213.42 mM; Need ultrasonic)
SMIL COCCNCCC1=CC(NC2=NC=C3CC(C4=CC=CC=C4F)C5=CC=CC=C5C3=N2)=CC=C1
Molekylvægt (g/mol) 468.57
Formel vægt 468.57
Procent renhed 99.38%
Grad Research
Fysisk form Solid