CAS RN 2301866-59-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
DO-264, MedChemExpress
MedChemExpress DO-264 is a selective and in vivo-active inhibitor of Abhydrolase Domain Containing 12 (ABHD12), with an IC50 of 11 nM.