CAS RN 229621-20-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
3,4-Bis(bromomethyl)-2,5-dihydro-2,2,5,5-tetramethyl-1H-pyrrol-1-yloxy Radical, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,4-Bis(bromomethyl)-2,5-dihydro-2,2,5,5-tetramethyl-1H-pyrrol-1-yloxy Radical, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,4-Bis(bromomethyl)-2,5-dihydro-2,2,5,5-tetramethyl-1H-pyrrol-1-yloxy Radical, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.