CAS RN 2174919-79-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
n8-Nor-17,17-dimethyl-5a-androst-13-en-2,3a-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
18-Nor-17,17-dimethyl-5a-androst-13-en-2,3a-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
18-Nor-17,17-dimethyl-5a-androst-13-en-2,3a-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.