CAS RN 215174-50-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Rimacalib, TRC
CAS: 215174-50-8 Molekylær formel: C22H23FN4O2 Molekylvægt (g/mol): 394.45 Synonym: SMP 114,N-[3-[(1S)-1-(2-Fluoro[1,1'-biphenyl]-4-yl)ethyl]-5-isoxazolyl]-4-morpholinecarboximidamide IUPAC navn: N-[3-[(1S)-1-(3-fluoro-4-phenylphenyl)ethyl]-1,2-oxazol-5-yl]morpholine-4-carboximidamide SMIL: C[C@@H](c1ccc(c(F)c1)c2ccccc2)c3cc(NC(=N)N4CCOCC4)on3
2
Rimacalib, MedChemExpress
MedChemExpress Rimacalib (SMP 114) is a Ca2+/calmodulin-dependent protein kinase II (CaMKII) inhibitor, with IC50s of ∼1 μM for CaMKIIα to ∼30 μM for CaMKIIγ.