CAS RN 2146095-85-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ SHIN 1
Potent SHMT inhibitor
2
SHIN1, MedChemExpress
MedChemExpress SHIN1 (RZ-2994) is a human serine hydroxymethyltransferse 1 and 2 (SHMT1/2) inhibitor with IC50s of 5 and 13 nM, respectively, in an in vitro assay.