CAS RN 21215-62-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ Calcitonin (human)
Endogenous calcitonin receptor agonist; inhibits bone resorption
2
Tocris Bioscience™ Calcitonin (human)
Endogenous calcitonin receptor agonist; inhibits bone resorption