Få mere at vide
Tocris Bioscience™ Calcitonin (human)
Endogenous calcitonin receptor agonist; inhibits bone resorption
Brand: Tocris Bioscience™ 6031/500U
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Calcitonin (human) is an endogenous calcitonin receptor agonist. Lowers systemic blood calcium levels and inhibits bone resorption.Tekniske data
| 95% | |
| Store at −20°C | |
| Yes | |
| Peptide | |
| CGNLSTCMLGTYTQDFNKFHTFPQTAIGVGAP |
| 21215-62-3 | |
| Endogenous calcitonin receptor agonist. | |
| 3417.87 | |
| 500 μg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only.