CAS RN 208848-19-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
5
af
5
Resultater
1
Tocris Bioscience™ ONO 6818
High affinity and selective human neutrophil elastase 1 (HNE1) inhibitor; orally active
2
Freselestat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Freselestat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Freselestat, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Tocris Bioscience™ ONO 6818
High affinity and selective human neutrophil elastase 1 (HNE1) inhibitor; orally active