Få mere at vide
Tocris Bioscience™ ONO 6818
High affinity and selective human neutrophil elastase 1 (HNE1) inhibitor; orally active
Brand: Tocris Bioscience™ 5651/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ONO 6818 is a high affinity and selective human neutrophil elastase 1 (HNE1) inhibitor (Ki = 12 nM). Exhibits >100-fold less activity at other related proteases including trypsin, pancreatic elastase, collagenase and murine macrophage elastase. Inhibits HNE-induced lung hemorrhaging and suppresses leukocyte levels in an in vivo model of COPD. Orally active.Tekniske data
| 5-Amino-N-[1-[[5-(1,1-dimethylethyl)-1,3,4-oxadiazol-2-yl]carbonyl]-2-methylpropyl]-6-oxo-2-phenyl-1(6H)-pyrimidineacetamide | |
| 208848-19-5 | |
| 10 mg |