CAS RN 208465-21-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Mesosulfuron-methyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Mesosulfuron-methyl, TRC
CAS: 208465-21-8 Molekylær formel: C17 H21 N5 O9 S2 Molekylvægt (g/mol): 503.51 Synonym: AE-F 130060,AE-F 130060-00,Mesomaxx,Mesosulfuron-methyl,Shima IUPAC navn: methyl 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-4-(methanesulfonamidomethyl)benzoate SMIL: COC(=O)c1ccc(CNS(=O)(=O)C)cc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2