CAS RN 2070018-31-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
CARM1-IN-1 hydrochloride, MedChemExpress
MedChemExpress CARM1-IN-1 hydrochloride is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM; shows very low activity against PRMT1 and SET7(IC50 > 600 uM).