CAS RN 205826-86-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(R)-alpha-[(2,2-Dimethyl-1-oxopropyl)amino]-4-hydroxybenzeneacetic Acid, TRC
CAS: 205826-86-4 Molekylær formel: C13 H17 N O4 Molekylvægt (g/mol): 251.28 Synonym: (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid,Amoxicillin Trihydrate Imp. H (EP) IUPAC navn: (2R)-2-(2,2-dimethylpropanoylamino)-2-(4-hydroxyphenyl)acetic acid SMIL: CC(C)(C)C(=O)N[C@@H](C(=O)O)c1ccc(O)cc1
2
(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.