Få mere at vide
(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid, Mikromol™
Artikelnummer. 30049940
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049940 25 mg
17990546 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049940 Leverandør Mikromol Leverandørnr. MM0232.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid
CAS 205826-86-4
Analyt- eller komponentnavne (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C13 H17 N O4
Certificeringer/compliance ISO 17025
Synonym Amoxicillin Trihydrate Imp. H (EP), Amoxicillin Imp. H (EP), Amoxicillin USP Related Compound H, Amoxicillin USP RC H, (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-(4-hydroxyphenyl)acetic Acid, Amoxicillin Related Compound H, Amoxicillin Trihydrate Impurity H, Amoxicillin Impurity H
InChI formel InChI=1S/C13H17NO4/c1-13(2,3)12(18)14-10(11(16)17)8-4-6-9(15)7-5-8/h4-7,10,15H,1-3H3,(H,14,18)(H,16,17)/t10-/m1/s1
SMIL CC(C)(C)C(=O)N[C@@H](C(=O)O)c1ccc(O)cc1
IUPAC navn (2R)-2-(2,2-dimethylpropanoylamino)-2-(4-hydroxyphenyl)acetic acid
Molekylvægt (g/mol) 251.28
Mængde 25 mg
Formel vægt 251.1158 g/mol