CAS RN 2055397-18-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ WM 8014
Potent and selective KAT6A and KAT6B inhibitor
2
WM-8014, MedChemExpress
MedChemExpress WM-8014 is an inhibitor of MOZ, a member of histone acetyltransferases, with an IC50 of 55 nM.