CAS RN 2034-23-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
5-Chloro-2-benzimidazolinone, TRC
CAS: 2034-23-3 Molekylær formel: C7 H5 Cl N2 O Molekylvægt (g/mol): 168.58 Synonym: 5-Chloro-1,3-dihydro-2H-benzimidazol-2-one,5-Chloro-1H-benzo[d]imidazol-2(3H)-one IUPAC navn: 5-chloro-1,3-dihydrobenzimidazol-2-one SMIL: Clc1ccc2NC(=O)Nc2c1
2
5-Chloro-1,3-dihydrobenzimidazol-2-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.