CAS RN 2032-34-0
IUPAC Navn:
3,3-diethoxypropanenitrile
Synonymer:
3,3-diethoxypropanenitrile
Molekylvægt (g/mol):
143.19
Molekylformel:
C7H13NO2
InChi Key:
WBOXEOCWOCJQNK-UHFFFAOYSA-N
SMILES:
CCOC(CC#N)OCC
1
–
3
af
3
Resultater
1
3,3-Diethoxypropionitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,3-Diethoxypropionitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,3-Diethoxypropionitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.